Learn More
CD19 Monoclonal Antibody (SJ25-C1), PE-Cyanine7, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD1912
Code : 50
Additional Details : Weight : 0.50000kg
Description
The R-phycoerythrin (PE)-Cy7® tandem conjugate permits simultaneous multicolor labeling and detection of multiple targets with excitation by a 488 nm laser without occupying traditional channels associated with FITC and PE. Alternatively, the PE dye tandem can be excited using a 532 nm or 561 nm laser.
CD19 is a member of the immunoglobulin superfamily and has two Ig like domains. The CD19 molecule is expressed on 100% of the peripheral B cells as defined by expression of kappa or lambda light chains. CD19 appears to be expressed on myeloid leukemia cells, particularly those of monocytic lineage. Leukemia phenotype studies have demonstrated that the earliest and broadest B cell restricted antigen is the CD19 antigen. The receptor for CD19 is an important functional regulator of normal and malignant B cell proliferation, and is expressed in all B cell precursor leukemias. Lymphocytes proliferate and differentiate in response to various concentrations of different antigens. The ability of the B cell to respond in a specific, yet sensitive manner to the various antigens is achieved with the use of low-affinity antigen receptors. CD19 is a cell surface molecule which assembles with the antigen receptor of B lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation. Besides being a signal-amplifying coreceptor for the B cell receptor (BCR), CD19 can also signal independently of BCR co-ligation and is a central regulatory component upon which multiple signaling pathways converge. Mutation of the CD19 gene results in hypogammaglobulinemia, whereas CD19 overexpression causes B cell hyperactivity.Specifications
CD19 | |
Monoclonal | |
PE-Cyanine7 | |
Cd19 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
Mouse | |
purified | |
ASR | |
930 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
SJ25-C1 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P15391 | |
Cd19 | |
Nalm 1 Human cell line | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |