missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse, FITC, Clone: SJ25C1, enQuireBio™
Mouse Monoclonal Antibody
£118.00 - £1040.00
Specifications
Antigen | CD19 |
---|---|
Clone | SJ25C1 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961778
|
EnquireBio
QAB29-F-25Tests |
25 Tests |
£118.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15951778
|
EnquireBio
QAB29-F-100Tests |
100 Tests |
£246.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15971778
|
EnquireBio
QAB29-F-500Tests |
500 Tests |
£1040.00
500 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD19 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
930 | |
IgG1 κ | |
Protein A/G Purified |
SJ25C1 | |
Flow Cytometry | |
FITC | |
RUO | |
NP_001171569 | |
CD19 | |
Primary | |
Store at 2-4°C. |