Learn More
CD20 Rabbit Polyclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments with Epredia™ CD20, Rabbit Polyclonal Antibody.
Brand: Epredia RB-9013-R7
Code : LD
Additional Details : Weight : 0.01200kg
Description
CD20 is a 33kDa leukocyte surface antigen consisting of four transmembrane regions and cytoplasmic N- and C-termini. CD20 is expressed primarily on B cells but has also been detected on both normal and neoplastic T cells. Activation of CD20 significantly increases DNA synthesis and is thought to involve basic helix-loop-helix leucine zipper transcription factors.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: C-terminus
Immunogen: Synthetic peptide derived from C-terminus of human CD20 protein
Molecular Weight: 33-37kDa
Positive Control: Tonsil
Cellular Localization: Membrane/cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
CD20 | |
Polyclonal | |
Unconjugated | |
Synthetic peptide derived from C-terminus of human CD20 protein | |
IVD | |
Primary | |
Prediluted |
Immunoprecipitation, Western Blot | |
Polyclonal | |
Rabbit | |
7 mL | |
B-Cell | |
Human | |
IgG |