missing translation for 'onlineSavingsMsg'
Learn More

CD20 Rabbit Polyclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments with Epredia™ CD20, Rabbit Polyclonal Antibody.

Brand:  Epredia RB-9013-P

Code : LD

Additional Details : Weight : 0.00250kg

 View more versions of this product

Product Code. 12663297

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

CD20 is a 33kDa leukocyte surface antigen consisting of four transmembrane regions and cytoplasmic N- and C-termini. CD20 is expressed primarily on B cells but has also been detected on both normal and neoplastic T cells. Activation of CD20 significantly increases DNA synthesis and is thought to involve basic helix-loop-helix leucine zipper transcription factors.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: C-terminus

Immunogen: Synthetic peptide derived from C-terminus of human CD20 protein

Molecular Weight: 33-37kDa

Positive Control: Tonsil

Cellular Localization: Membrane/cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation
Specifications

Specifications

CD20
Polyclonal
200µg/mL
Rabbit
1 mL
B-Cell
Human
IgG
Immunoprecipitation, Western Blot
Polyclonal
Unconjugated
Synthetic peptide derived from C-terminus of human CD20 protein
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers