Learn More
CD209 (DC-SIGN) Monoclonal Antibody (eB-h209), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£246.00
Specifications
Antigen | CD209 (DC-SIGN) |
---|---|
Clone | eB-h209 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Description: The eB-h209 monoclonal antibody reacts with human CD209, also known as DC-SIGN, a 44 kDa type II transmembrane protein. DC-SIGN contains a C-type lectin binding domain and binds ICAM-3, ICAM-2, and HIV virus. Human dendritic cells preferentially express DC-SIGN. It has been postulated that DC-SIGN serves as a receptor for capture, trafficking, and transmission of HIV to T cells and supports primary immune response. eB-h209 was developed against a C-terminal peptide of human DC-SIGN. Applications Reported: The eB-h209 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eB-h209 antibody has been pre-titrated and tested by flow cytometric analysis of cultured human dendritic cells and peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD209 (DC-SIGN) Monoclonal antibody specifically detects CD209 (DC-SIGN) in Human samples. It is validated for Flow Cytometry.Specifications
CD209 (DC-SIGN) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q9NNX6 | |
CD209 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD209 |
eB-h209 | |
Flow Cytometry | |
APC | |
Rat | |
Human | |
30835 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.