missing translation for 'onlineSavingsMsg'
Learn More

CD215 (IL-15Ra) Monoclonal Antibody (eBioJM7A4), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen 14-7159-80

 View more versions of this product

Product Code. 15278027

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Description: The eBioJM7A4 monoclonal antibody reacts with the IL-15 receptor alpha chain (IL-15Ralpha). IL-15 and IL-2 are cytokines with overlapping, but distinct, biologic effects. Their receptors share 2 subunits, the IL-2Rbeta and gamma chains, which are essential for signal transduction. The IL-15Ralpha receptor is structurally related to IL-2Ralpha. However, the IL-15Ralpha alone binds IL-15 with a 1,000-fold higher affinity than that seen with IL-2Ralpha and IL-2. In the human, 3 differentially spliced human IL-15Ralpha variants that are all capable of high affinity binding of IL-15 have been isolated. The cytoplasmic domain of IL-15Ralpha, like that of IL-2Ralpha, is dispensable for mitogenic signaling, suggesting that the primary role of the alpha chains is to confer high affinity binding. At high concentrations, IL-15, like IL-2, is able to signal through a complex of IL-2Rbeta and IL-2Rgamma in the absence of the alpha subunit. Applications Reported: This eBioJM7A4 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunoblotting (WB). Applications Tested: This eBioJM7A4 antibody has been tested by flow cytometric analysis of LPS and IFN gamma-stimulated normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.

Interleukin 15 (IL-15RA) is a cytokine receptor that specifically binds IL-15 with high affinity. IL-15 regulates T and natural killer cell activation and proliferation. The IL-15 and IL-2 receptors share two subunits, IL-2R beta and IL-2R gamma, and IL-15RA is structurally related to IL-2RA, an additional IL-2-specific alpha subunit necessary for high affinity IL-2 binding. Unlike IL-2RA, IL-15RA is capable of binding IL-15 with high affinity independent of the other subunits. IL-15RA is thought to enhance cell proliferation and expression of apoptosis inhibitor Bcl-xL and Bcl-2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD215 (IL-15Ra)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q13261
IL15RA
Affinity chromatography
RUO
3601
4° C
Liquid
Flow Cytometry, Immunoprecipitation, Western Blot
eBioJM7A4
Unconjugated
IL15RA
AA690181; CD215; IL 15 receptor; IL 15 receptor subunit a; IL 15R; IL 15R alpha; IL 15R a; IL15 receptor; IL15 receptor subunit alpha; IL-15 receptor subunit alpha; IL15 receptor subunit a; IL15R; IL15R alpha; IL15R a; Il15ra; IL-15RA; IL-15R-alpha; Interleukin 15 receptor; interleukin 15 receptor alpha isoform EM2; interleukin 15 receptor alpha isoform IC2; interleukin 15 receptor alpha isoform IC3; interleukin 15 receptor alpha isoform IC4; interleukin 15 receptor alpha isoform IC5; interleukin 15 receptor alpha isoform IC6; interleukin 15 receptor alpha isoform IC7; interleukin 15 receptor alpha isoform IC8; interleukin 15 receptor subunit alpha; Interleukin 15 receptor subunit a; interleukin 15 receptor, alpha; interleukin 15 receptor, alpha chain; interleukin-15 receptor subunit alpha; sIL 15R; sIL-15 receptor subunit alpha; sIL15R; sIL-15RA; sIL-15R-alpha; soluble IL 15R; soluble IL15R; Soluble interleukin-15 receptor subunit alpha
Mouse
25 μg
Primary
Human
Antibody
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.