missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD23 Mouse, PE, Clone: EBVCS-5, enQuireBio™
Mouse Monoclonal Antibody
£136.00 - £1375.00
Specifications
Antigen | CD23 |
---|---|
Clone | EBVCS-5 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961848
|
EnquireBio
QAB32-PE-25Tests |
25 Tests |
£136.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15951848
|
EnquireBio
QAB32-PE-100Tests |
100 Tests |
£311.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15971848
|
EnquireBio
QAB32-PE-500Tests |
500 Tests |
£1375.00
500 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD23 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
2208 | |
IgG1 κ | |
Protein A/G Purified |
EBVCS-5 | |
Flow Cytometry | |
PE | |
RUO | |
NP_001993 | |
FCER2 | |
Primary | |
Store at 2-4°C. |