missing translation for 'onlineSavingsMsg'
Learn More

CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), NovaFluor™ Yellow 570, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£136.00 - £302.00

Specifications

Antigen CD24
Clone eBioSN3 (SN3 A5-2H10)
Concentration 0.2 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17876611
View Documents
Invitrogen
H064T02Y01
25 Tests This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17836621
View Documents
Invitrogen
H064T03Y01
100 Tests This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), NovaFluor™ Yellow 570, eBioscience™, Invitrogen™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.