Learn More
CD24 Monoclonal Antibody (M1/69), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£81.50 - £205.00
Specifications
Antigen | CD24 |
---|---|
Clone | M1/69 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry, Functional Assay, Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15216867
|
Invitrogen
14-0242-82 |
100 μg |
£81.50
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15226867
|
Invitrogen
14-0242-85 |
500 μg |
£205.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The M1/69 monoclonal antibody reacts with the mouse CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50 kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. CD24 is a variably glycosylated molecule resulting in heterogeneity of molecular mass of this antigen on cells of different lineages and antibodies to CD24 exhibit subtle differences in staining level on lymphocyte populations. The expression of CD24 has been used to resolve stages of B lymphopoiesis in mouse bone marrow. It has been reported that P-selectin (CD62P) binds to CD24. Applications Reported: The M1/69 antibody has been reported for use in flow cytometric analysis, immunoblotting (WB), and immunohistochemical staining. M1/69 has also been reported in in vitro functional assays. (Please use Functional Grade purified M1/69, cat. 16-0242, in functional assays.). Applications Tested: The M1/69 antibody has been tested by flow cytometric analysis of mouse splenocytes and thymocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.Specifications
CD24 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD24; CD24 antigen; CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24 molecule; CD24A; CD24a antigen; cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; heat-stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; Nectadrin; nectadrin heat stable antigen; R13-Ag; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen; X62 heat stable antigen | |
Cd24a | |
Primary | |
4° C | |
Cd24a |
M1/69 | |
Flow Cytometry, Functional Assay, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P24807 | |
12484 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.