Learn More
CD25 Monoclonal Antibody (PC61.5), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£45.71 - £304.00
Specifications
Antigen | CD25 |
---|---|
Clone | PC61.5 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15259499
|
Invitrogen
13-0251-81 |
50 μg | |||||||
15269499
|
Invitrogen
13-0251-82 |
100 μg |
£90.75
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15279499
|
Invitrogen
13-0251-85 |
500 μg |
£183.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15289499
|
Invitrogen
13-0251-86 |
1 mg |
£304.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The PC61.5 antibody reacts with mouse CD25, the 55 kDa interleukin-2 receptor alpha chain (IL-2R alpha). CD25 is expressed by early progenitors of the T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) to form the high affinity IL-2 receptor. Binding of IL-2 to both the high and low affinity classes of IL-2 receptor is inhibited by the PC61.5 antibody. CD25 plays a role in lymphocyte differentiation and activation/proliferation. Applications Reported: The PC61.5 antibody has been reported for use in flow cytometric analysis. Applications Tested: The PC61.5 antibody has been tested by flow cytometric analysis of mouse splenocytes and Con A-stimulated splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
CD25 (IL2 receptor alpha chain/IL2RA) is a cytokine that plays a role in the proliferation of T and B lymphocytes. The receptor of this cytokine (IL2RA) is a heterotrimeric protein complex with a gamma chain also shared by interleukin 4 (IL4) and interleukin 7 (IL7). IL2RA, IL2R beta chain (IL2RB), and the IL2R gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric IL2RA chains result in low-affinity receptor, while homodimeric IL2RB chains produce a medium-affinity receptor. The expression of IL2 in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. IL2 is primarily produced by mature T cells. IL2 plays an important role as a growth factor, differentiation factor, and regulator of cell death. IL-2 stimulates the proliferation of B cells, augments natural killer cell activity, and inhibits granulocyte macrophage colony formation. The targeted disruption of a similar gene in mice leads to μgcerative colitis-like disease, which suggests a role in the immune response to antigenic stimuli. Mutations in this gene are associated with interleukin 2 receptor alpha deficiency.Specifications
CD25 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P01590 | |
IgG1 λ | |
Affinity chromatography | |
Antibody |
PC61.5 | |
Flow Cytometry | |
Biotin | |
Rat | |
Mouse | |
16184 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.