missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Rat, PerCP-Cy5.5, Clone: PC61.5, enQuireBio™
Rat Monoclonal Antibody
£99.15 - £256.00
Specifications
Antigen | CD25 |
---|---|
Clone | PC61.5 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15951868
|
EnquireBio
QAB34-PCP55-25ug |
25 μg |
£99.15
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15941868
|
EnquireBio
QAB34-PCP55-100ug |
100 μg |
£256.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD25 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
16184 | |
IgG1 λ | |
Protein A/G Purified |
PC61.5 | |
Flow Cytometry | |
PerCP-Cyanine5.5 | |
RUO | |
NP_032393 | |
Il2ra | |
Primary | |
Store at 2-4°C. |