missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, Clone: 4AT3, Abnova™

Mouse monoclonal antibody raised against human CD3.

Brand:  Abnova MAB12366.100uL

Product Code. 16188749

  • £328.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

CD3
Monoclonal
Unconjugated
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Mouse
Protein A/G affinity chromatography
RUO
Human
Liquid
Flow Cytometry, Immunofluorescence
4AT3
Mouse monoclonal antibody raised against human CD3.
In PBS, pH 7.4 (0.09% sodium azide)
CD3
100 μL
Primary
Store at 4°C. Do not freeze.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.