Learn More
CD3 Mouse, PE-Cy7, Clone: SK7, enQuireBio™
Mouse Monoclonal Antibody
Brand:  EnquireBio QAB4-PE7-25Tests
 View more versions of this product
Product Code. 15941958
  • £182.00 / 25 tests
Explore more special offers
Specifications
CD3
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD3E
Protein A/G Purified
RUO
916
IgG1 κ
Flow Cytometry
SK7
PE-Cyanine7
NP_000724
Mouse
25 Tests
Primary
Store at 2-4°C.