Learn More
CD339 (Jagged 1) Monoclonal Antibody (HMJ1-29), PE, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen 12-3391-82
Description
Description: This HMJ1-29 monoclonal antibody reacts with mouse Jagged-1, one of five type I transmembrane proteins that serves as a Notch receptor ligand. Upon binding the Notch receptor (e.g. Notch 1-4), Jagged-1 undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases an intracellular fragment whose biological function remains controversial. Jagged-1 is expressed by thymic lymphoid and stromal cells, as well as macrophages and CD8+CD11c+ dendritic cells in the spleen. The HMJ1-29 antibody is reported to crossreact to rat. Applications Reported: This HMJ1-29 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HMJ1-29 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.Specifications
CD339 (Jagged 1) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q63722, Q9QXX0 | |
JAG1 | |
Affinity chromatography | |
RUO | |
16449, 29146 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
HMJ1-29 | |
PE | |
JAG1 | |
ABE2; AGS; AHD; AWS; CD339; Gsfabe2; HJ1; Htu; JAG1; jagged 1; jagged canonical Notch ligand 1; Jagged1; JAGL1; Ozz; protein jagged-1; Ser-1; Serrate-1 | |
Armenian Hamster | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
For Research Use Only.