Learn More
CD4 Monoclonal Antibody (RM4-4), FITC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£83.75 - £202.00
Specifications
Antigen | CD4 |
---|---|
Clone | RM4-4 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15257999
|
Invitrogen
11-0043-82 |
100 μg |
£83.75
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15267999
|
Invitrogen
11-0043-85 |
500 μg |
£202.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: The RM4-4 monoclonal antibody reacts with the mouse CD4 molecule, a 55 kDa cell surface receptor expressed by the majority of thymocytes, a subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with the protein tyrosine kinase lck through its cytoplasmic tail. Binding of RM4-4 does not block binding of the CD4 monoclonal antibodies RM4-5 or GK1.5. Applications Reported: This RM4-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RM4-4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].Specifications
CD4 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen | |
CD4 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CD4 |
RM4-4 | |
Flow Cytometry | |
FITC | |
Rat | |
Mouse | |
P06332 | |
12504 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.