missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat anti-Mouse, FITC, Clone: 4AM15, Abnova™
Rat monoclonal antibody raised against mouse Cd4.
Brand: Abnova MAB12417.50Reactions
Additional Details : Weight : 0.00010kg
Specifications
CD4 | |
Monoclonal | |
FITC | |
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user. | |
L3T4/Ly-4 | |
Rat | |
Protein A/G affinity chromatography | |
RUO | |
12504 | |
Store at 4°C. Do not freeze. | |
IgG2b |
Flow Cytometry, Immunofluorescence | |
4AM15 | |
Rat monoclonal antibody raised against mouse Cd4. | |
In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA) | |
Cd4 | |
Cd4 | |
50 Reactions | |
Primary | |
Mouse | |
Liquid |