Learn More
CD45 Rabbit anti-Human, Unconjugated, Clone: 6O19, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80297-1-RR-100UL
This item is not returnable.
View return policy
Description
The CD45 antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human CD45. This antibody recognizes human, rat antigen. The CD45 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
CD45 | |
6O19 | |
Rabbit | |
Primary | |
Human, Rat |
Western Blot, Immunohistochemistry, Flow Cytometry | |
Unconjugated | |
100 μL | |
5788 | |
IgG |