missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RB Antibody (PD7/26), Novus Biologicals™
Mouse Monoclonal Antibody
£215.00 - £372.00
Specifications
Antigen | CD45RB |
---|---|
Clone | PD7/26 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 1-2 ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 0.25-0.5 ug/mL |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18403442
|
Novus Biologicals
NBP2-32937-0.1mg |
0.1 mg |
£372.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18460652
|
Novus Biologicals
|
0.02 mg |
£215.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45RB Monoclonal antibody specifically detects CD45RB in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
CD45RB | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Human peripheral blood lymphocytes maintained in T cell growth factor | |
Primary | |
Store at 4C. |
PD7/26 | |
Western Blot 1-2 ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 0.25-0.5 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P08575, P08575 | |
5788 | |
IgG1 κ | |
Protein A or G purified |