missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CD47 (Human) Recombinant Protein

Human CD47 full-length ORF (NP_001768.1) recombinant protein without tag.

Brand:  Abnova™ H00000961-G01.2ug

Product Code. 16185944

  • £377.00 / 2µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sequence: MWPLVAALLLGSACCGSAQLLFNKTKSVEFTFCNDTVVIPCFVTNMEAQNTTEVYVKWKFKGRDIYTFDGALNKSTVPTDFSSAKIEVSQLLKGDASLKMDKSDAVSHTGNYTCEVTELTREGETIIELKYRVVSWFSPNENILIVIFPIFAILLFWGQFGIKTLKYRSGGMDEKTIALLVAGLVITVIVIVGAILFVPGEYSLKNATGLGLIVTSTGILILLHYYVFSTAIGLTSFVIAILVIQVIAYILAVVGLSLCIAACIPMHGPLLISGLSILALAQLLGLVYMKFVASNQKTIQPPRKAVEEPLNAFKESKGMMNDE
Specifications

Specifications

NP_001768.1
CD47 molecule
2 μg
IAP, MER6, OA3
Wheat Germ (in vitro)
Liquid
961
Wheat germ expression system
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD47
25 mM Tris-HCl of pH 8.0 containing 2% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.