missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD47 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547215
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 15% cross-reactivity with recombinant human CD47 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CD47 Polyclonal antibody specifically detects CD47 in Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
CD47 | |
Polyclonal | |
Unconjugated | |
CD47 | |
9130415E20Rik; AA407862; AI848868; antigen identified by monoclonal antibody 1D8; antigenic surface determinant protein OA3; AW108519; B430305P08Rik; Cd47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 antigen/integrin-associated protein; CD47 glycoprotein; CD47 molecule; CD47/IAP; IAP; integrin associated protein; integrin-associated protein; integrin-associated signal transducer; Itgp; Leukocyte surface antigen CD47; MER6; OA3; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
16423, 29364 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P97829, Q61735 | |
CD47 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD47 N-terminal IgV-like Gln19-Pro158 | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |