missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Mouse, FITC, Clone: L17F12, enQuireBio™
Mouse Monoclonal Antibody
£109.00 - £1100.00
Specifications
Antigen | CD5 |
---|---|
Clone | L17F12 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931538
|
EnquireBio
QAB14-F-25Tests |
25 Tests |
£109.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15921538
|
EnquireBio
QAB14-F-100Tests |
100 Tests |
£256.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15941538
|
EnquireBio
QAB14-F-500Tests |
500 Tests |
£1100.00
500 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD5 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
921 | |
IgG2a κ | |
Protein A/G Purified |
L17F12 | |
Flow Cytometry | |
FITC | |
RUO | |
NP_055022 | |
CD5 | |
Primary | |
Store at 2-4°C. |