missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Rat, APC, Clone: 53-7.3, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB13-APC-25ug
This item is not returnable.
View return policy
Specifications
CD5 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Cd5 | |
Protein A/G Purified | |
RUO | |
12507 | |
IgG2a κ |
Flow Cytometry | |
53-7.3 | |
APC | |
XP_006526684 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |