missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD53 Mouse anti-Human, Clone: HD77, Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £432.00
Specifications
Antigen | CD53 |
---|---|
Clone | HD77 |
Concentration | 1 mg/ml |
Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18321092
|
Bio-Techne
NBP3-12058-0.025MG |
0.025 mg |
£195.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18371712
|
Bio-Techne
NBP3-12058-0.2MG |
0.2 mg |
£432.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD53 Monoclonal antibody specifically detects CD53 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
CD53 | |
1 mg/ml | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Immunology | |
PBS | |
963 | |
IgG1 κ | |
Protein A purified |
HD77 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
Monoclonal | |
Purified | |
RUO | |
Human | |
antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53 | |
This antibody was raised by immunising mice with human leukemia cells. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title