missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD59 Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
Brand: Novus Biologicals™ NBP2-79401-15PLATES
This item is not returnable.
View return policy
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD59.Specifications
234.38 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein | |
CD59 | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 15000 pg/mL is recommended. |
234.38-15000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD59. | |
Sandwich ELISA | |
966 | |
Rabbit | |
15 Plates | |
Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers | |
CD59 |