missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD61 (Integrin beta 3) Monoclonal Antibody (2C9.G3), NovaFluor™ Yellow 570, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
£161.00 - £303.00
Specifications
Antigen | CD61 (Integrin beta 3) |
---|---|
Clone | 2C9.G3 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17836236
|
Invitrogen
M053T02Y01 |
25 μg |
£161.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17876206
|
Invitrogen
M053T03Y01 |
100 μg |
£303.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD61 (GPIIIa, ITGB3) is a glycoprotein found on megakaryocytes, platelets and their precursors. CD61 antigen plays a role in platelet aggregation and also as a receptor for fibrinogen, fibronectin, von Willebrand factor and vitronectrin. CD61 is a 105 kDa glycoprotein that associates with either the alpha 11b integrin (CD41) or the alpha V integrin (CD51) at the cell surface. CD61 is expressed on platelets and megakaryocytes in association with CD41, and on endothelial cells, monocytes, platelets and osteoclasts in association with CD51. CD61 is a receptor for fibrinogen, fibronectin, Von Willebrand Factor, vitronectin and thrombospondin. The CD61 protein product is composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. CD61 is found along with the alpha IIb chain in platelets. Like other integrins, CD61 is known to participate in cell adhesion as well as cell-surface mediated signaling. Diseases associated with CD61 dysfunction include Glanzmann Thrombasthenia and Platelet type-16 Bleeding Disorder.Specifications
CD61 (Integrin beta 3) | |
0.1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
antigen CD61; BDPLT16; BDPLT2; beta3 integrin; CD61; EGK_08849; glycoprotein GPIIIa; glycoprotein IIIa; GP3A; GPIIIa; GT; INGRB3; integrin beta 3; integrin beta 3 (Cd61); integrin beta-3; integrin beta-3 subunit; integrin subunit beta 3; integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGB3; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet gpIIIa; platelet membrane glycoprotein IIIa | |
ITGB3 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
2C9.G3 | |
Flow Cytometry | |
NovaFluor Yellow 570 | |
Armenian Hamster | |
Mouse, Rat | |
O54890 | |
16416, 29302 | |
IgG | |
Affinity chromatography | |
ITGB3 |