missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD71 Rat, anti-Mouse, Clone: RI7217, Proteintech
Rat Monoclonal Antibody
£39.00 - £104.00
Specifications
| Antigen | CD71 |
|---|---|
| Clone | RI7217 |
| Concentration | 0.5 mg/ml |
| Applications | Flow Cytometry |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18069415
|
Proteintech
65289-1-IG-500UG |
500 μg |
£104.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18008935
|
Proteintech
65289-1-IG-100UG |
100 μg |
£39.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD71 antibody from Proteintech is a rat monoclonal antibody generated with dmso induced friend erythroleukemia 745.6 of mouse CD71. This antibody recognizes mouse antigen. The CD71 antibody has been validated for the following applications: FC analysis.
Specifications
| CD71 | |
| 0.5 mg/ml | |
| Liquid | |
| Mouse | |
| 2610028K12Rik, AI195355, AI426448, AU015758, CD71, E430033M20Rik, Mtvr 1, Mtvr1, p90, TFR, TFR1, Tfrc, transferrin receptor, Trfr | |
| Primary |
| RI7217 | |
| Flow Cytometry | |
| Rat | |
| 22042 | |
| IgG2a κ | |
| Store at 2°C to 8°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title