missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 beta Monoclonal Antibody (CT-CD8b), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517843
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17843 targets CD8b/LYT3 in FACS applications and shows reactivity with Mouse samples. The MA5-17843 immunogen is mouse CD8 β.
The CD8B antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen, acting as a coreceptor, and the T-cell receptor on the T lymphocyte recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The functional coreceptor is either a homodimer composed of two alpha chains, or a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. This gene encodes the CD8 beta chain isoforms. Multiple alternatively spliced transcript variants encoding distinct membrane associated or secreted isoforms have been described. A pseudogene, also located on chromosome 2, has been identified.Specifications
CD8 beta | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P10300 | |
Cd8b1 | |
Mouse CD8 β | |
200 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
CT-CD8b | |
Unconjugated | |
Cd8b1 | |
CD8; CD8 antigen 37 kDa chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8a; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; CD8B1; CD8beta; CD8BP; LEU2; Leu-2; LY3; Ly-3; Ly-C; Lymphocyte antigen 3; Lyt3; Lyt-3; MAL; OX-8 membrane antigen; p32; P37; T lymphocyte surface glycoprotein beta chain; T-cell membrane glycoprotein Ly-3; T-cell surface glycoprotein; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-3 | |
Rat | |
Protein G | |
RUO | |
12526 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |