missing translation for 'onlineSavingsMsg'
Learn More

CD80 Armenian Hamster, PE, Clone: 16-10A1, enQuireBio™

Armenian Hamster Monoclonal Antibody

Brand:  EnquireBio QAB50-PE-25ug

 View more versions of this product

Product Code. 15972148

  • £71.60 / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

CD80
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Cd80
Protein A/G Purified
RUO
12519
IgG
Flow Cytometry
16-10A1
PE
XP_017172347
Armenian Hamster
25 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers