Learn More
CD80 (B7-1) Armenian Hamster anti-Mouse, FITC, Clone: 16-10A1, Proteintech
Armenian Hamster Monoclonal Antibody
Brand: Proteintech FITC-65076-100UG
This item is not returnable.
View return policy
Description
The CD80 (B7-1) antibody from Proteintech is a armenian hamster monoclonal antibody generated with cho cell line transfected with mouse b7 (cd80) of mouse CD80 (B7-1). This antibody recognizes mouse antigen. The CD80 (B7-1) antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
CD80 (B7-1) | |
Monoclonal | |
0.5 mg/mL | |
Armenian Hamster | |
Primary | |
Mouse |
Flow Cytometry | |
16-10A1 | |
FITC | |
100 μg | |
12519 | |
IgG |