missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD84 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12566-1-AP-150UL
This item is not returnable.
View return policy
Description
CD84 is expressed on mature B cells and on B cell lines, including pre B cell lines, but not on plasma cell lines. Immunohistochemical studies demonstrated that CD84 strongly expressed on tissues macrophages.Specifications
CD84 | |
Polyclonal | |
Unconjugated | |
CD84 | |
CD84, CD84 molecule, Cell surface antigen MAX.3, hCD84, Hly9 beta, LY9B, mCD84, SLAM family member 5, SLAMF5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12523, 8832 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q18PI6, Q9UIB8 | |
Cd84 | |
CD84 Fusion Protein Ag3192 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD84 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction