Learn More
CD95 (APO-1/Fas) Monoclonal Antibody (EOS9.1), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£2595.00 - £12950.00
Specifications
Antigen | CD95 (APO-1/Fas) |
---|---|
Clone | EOS9.1 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Functional Assay |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17121102
|
Affymetrix eBioscience
16-0958-38 |
5 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17101102
|
Affymetrix eBioscience
16-0958-025 |
25 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17111102
|
Affymetrix eBioscience
16-0958-050 |
50 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The EOS9.1 monoclonal antibody reacts with human CD95 (Fas, Apo-1), a 40-50 kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, activated lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. EOS9.1 does not block binding of DX2, another antibody specific for human CD95. Applications Reported: The EOS9.1 antibody has been reported for use in flow cytometric analysis. EOS9.1 is also effective in inducing apoptosis in in vitro functional studies. Applications Tested: The EOS9.1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/ μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD95 (APO-1/Fas) Monoclonal antibody specifically detects CD95 (APO-1/Fas) in Human samples. It is validated for Flow Cytometry, Functional AssaySpecifications
CD95 (APO-1/Fas) | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
AI196731; ALPS1A; APO1; APO-1; Apo1 antigen; apo-1 antigen; APO-1 cell surface antigen; Apo-1 Fas; apoptosis antigen 1; apoptosis signaling receptor FAS; apoptosis-mediating surface antigen FAS; APT1; CD95; CD95 antigen; fas; Fas (TNF receptor superfamily member 6); Fas (TNF receptor superfamily member); Fas (TNF receptor superfamily, member 6); Fas AMA; Fas antigen; Fas antigen (ATP1); Fas cell surface death receptor; Fas receptor; FAS1; FASLG receptor; FASTM; lpr; lymphoproliferation; OTTHUMP00000020045; OTTHUMP00000020046; OTTHUMP00000020051; OTTHUMP00000059646; TNF receptor superfamily member 6; TNFR6; Tnfrsf6; Tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily, member 6 | |
IgM κ | |
Affinity chromatography | |
Antibody |
EOS9.1 | |
Flow Cytometry, Functional Assay | |
Functional Grade | |
Mouse | |
Human | |
P25445 | |
355 | |
Primary | |
4° C | |
FAS |