Learn More
CD98/SLC3A2 Mouse anti-Human, Unconjugated, Clone: 2B10F5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66883-1-IG-20UL
This item is not returnable.
View return policy
Description
The CD98/SLC3A2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD98/SLC3A2. This antibody recognizes human antigen. The CD98/SLC3A2 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
CD98/SLC3A2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
2B10F5 | |
Unconjugated | |
20 μL | |
6520 | |
IgG2a |