missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10155-1-AP-20UL
This item is not returnable.
View return policy
Description
This protein is a small GTPase of the Rho subfamily, which regulates signaling pathways that control diverse cellular functions including cell morphology, migration, endocytosis, and cell progression. This protein is highly similar to Saccharomyces cerevisiae Cdc 42, and is able to complement the yeast cdc42 1 mutant. The product of oncogene Dbl was reported to specifically catalyze the dissociation of GDP from this protein. This protein could regulate actin polymerization through its direct binding to Neural Wiskott-Aldrick syndrome protein (N-WASP), which subsequently activates Arp2/3 complex. Alternative splicing of this gene results in at least two transcript variants.Specifications
CDC42 | |
Polyclonal | |
Unconjugated | |
CDC42 | |
CDC42, CDC42Hs, G25K, G25K GTP binding protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12540, 64465, 998 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P60766, P60953, Q8CFN2 | |
Cdc42 | |
CDC42 Fusion Protein Ag0207 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction