missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA575302
Additional Details : Weight : 0.01000kg
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
F-box-like protein which is required for entry into mitosis. Acts by participating in E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase. [UniProt]Specifications
CDCA3 | |
Polyclonal | |
Unconjugated | |
CDCA3 | |
2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14793, 297594, 83461 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Q68FW2, Q99618, Q99M54 | |
CDCA3 | |
Synthetic peptide corresponding to amino acids 220-265 of Human CdcA3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |