missing translation for 'onlineSavingsMsg'
Learn More

CDCA3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA5110918

Product Code. 17128843

  • £502.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: KEEARQPTET PVASQSSDKP SRDPETPRSS GSMRNRWKPN SSKVLGRSPL TILQDDNSPG TLTLRQGKRP SPLSENVSEL KEGAILGTGR LLKTGGRAWE Q

F-box-like protein which is required for entry into mitosis. Acts by participating in E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase. [UniProt]
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CDCA3
Polyclonal
Unconjugated
CDCA3
2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1
Rabbit
Antigen affinity chromatography
RUO
83461
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q99618
CDCA3
Recombinant Protein corresonding to Human CDCA3
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers