missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5101298
Additional Details : Weight : 0.01000kg
Description
Antibody detects endogenous levels of total CDCA3.
F-box-like protein which is required for entry into mitosis. Acts by participating in E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase. [UniProt]Specifications
CDCA3 | |
Polyclonal | |
Unconjugated | |
CDCA3 | |
2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
14793, 297594, 83461 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q68FW2, Q99618, Q99M54 | |
CDCA3 | |
A synthesized peptide derived from human CDCA3(Accession Q99618), corresponding to amino acid residues L231-S268. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |