Learn More
CDCA3 Recombinant Rabbit Monoclonal Antibody (1J5L2), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA542866
Additional Details : Weight : 0.01000kg
Description
Positive test controls include: K-562. Immunogen sequence: MGSAKSVPVT PARPPPHNKH LARVADPRSP SAGILRTPIQ VESSPQPGLP AGEQLEGLKH AQDSDPRSPT LGIARTPMKT SSGDPPSPLV KQLSEVFETE
F-box-like protein which is required for entry into mitosis. Acts by participating in E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase. [UniProt]Specifications
CDCA3 | |
Recombinant Monoclonal | |
1.35 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68FW2, Q99618, Q99M54 | |
CDCA3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CDCA3 (Q99618). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1J5L2 | |
Unconjugated | |
CDCA3 | |
2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14793, 297594, 83461 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |