missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK4 Antibody Cocktail, Invitrogen™
Antibody Cocktail
Brand: Invitrogen MA513720
This item is not returnable.
View return policy
Description
MA5-13720 targets Cdk4 in IF, IP, and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-13720 immunogen is purified recombinant cdk4 protein.
CDK4/cyclin D3 serine/threonine kinase is dimeric protein complex. CDK4 is a member of the cyclin-dependent protein kinase family and is involved in the control of cell proliferation during the G1 phase of cell cycle. CDK4 forms a complex with the D-type cyclins and is inhibited by p16 (cyclin-dependent kinase inhibitor-2). CDK4 can mediate phosphorylation of the C-terminal region of Rb protein leading to an active transcriptional repression of E2F complex. CDC37 and HSP90 can preferentially associate with the fraction of CDK4 not bound to D-type cyclins. SMAD3 is a major physiologic substrate of the G1 cyclin-dependent kinases CDK4 and CDK2. Although CDK6 and CDK4 can both phosphorylate multiple residues in the Rb protein, they do so with different residue selectivity in vitro; CDK6 phosphorylates Thr821 while CDK4 phosphorylates Thr826 on Rb protein.
Specifications
CDK4 | |
Cocktail | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P11802, P30285, P35426 | |
Cdk4 | |
Purified recombinant cdk4 protein | |
500 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
DCS-31, DCS-35 | |
Unconjugated | |
Cdk4 | |
Cdk4; Cell division protein kinase 4; CMM3; Crk3; cyclin dependent kinase 4; cyclin-dependent kinase 4; MGC14458; p34<PSK-J3>/cdk4; PSK-J3; serine/threonine kinase; Unknown (protein for MGC:133903) | |
Mouse | |
Protein G | |
RUO | |
1019, 12567, 94201 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CDK4 Antibody Cocktail, Invitrogen™ > 500 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction