Learn More
Invitrogen™ Human CDK4/cyclin D3, GST Tag Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ A33736
 View more versions of this product
Product Code. 17188845
  • £7060.00 / 1mg
Explore more special offers
Description
Description
CDK4/cyclin D3 serine/threonine kinase is dimeric protein complex. CDK4 is a member of the cyclin-dependent protein kinase family and is involved in the control of cell proliferation during the G1 phase of cell cycle. CDK4 forms a complex with the D-type cyclins and is inhibited by p16 (cyclin-dependent kinase inhibitor-2). CDK4 can mediate phosphorylation of the C-terminal region of Rb protein leading to an active transcriptional repression of E2F complex. CDC37 and HSP90 can preferentially associate with the fraction of CDK4 not bound to D-type cyclins. SMAD3 is a major physiologic substrate of the G1 cyclin-dependent kinases CDK4 and CDK2. Although CDK6 and CDK4 can both phosphorylate multiple residues in the Rb protein, they do so with different residue selectivity in vitro; CDK6 phosphorylates Thr821 while CDK4 phosphorylates Thr826 on Rb protein.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P11802
Kinase Assay
1019
Human CDK4/cyclin D3, GST Tag
Cdk4; Cell division protein kinase 4; CMM3; Crk3; cyclin dependent kinase 4; cyclin-dependent kinase 4; MGC14458; PSK-J3; serine/threonine kinase; Unknown (protein for MGC:133903)
CDKs and CHKs
Human
GST-tag
-80°C, Avoid Freeze/Thaw Cycles
Insect cells
Liquid
See Label
50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5
60.0-58.8 kDa
1 mg
CDK4
Unconjugated
Recombinant
CDK4: 1-303, Cyclin D3: 1-292
Dry Ice
Full Length, Recombinant, Active