missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | CDK6 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16812524
|
Proteintech
14052-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16802524
|
Proteintech
14052-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IARS2 belongs to the class-I aminoacyl-tRNA synthetase family. Members of class I have two highly conserved sequence motifs. They aminoacylate at the 2′-OH of an adenosine nucleotide, and are usually monomeric or dimeric (one or two subunits, respectively). Both classes of aminoacyl-tRNA synthetases are multidomain proteins. The catalytic domains of all the aaRSs of a given class are found to be homologous to one another, while class I and class II aaRSs are unrelated to one another. The class I aaRSs have the ubiquitous Rossmann fold and have the antiparallel beta-strands architecture while the class II aaRSs have a unique fold made up of antiparallel beta-strands.Specifications
CDK6 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q00534, Q64261 | |
1021, 114483, 12571 | |
CDK6 Fusion Protein Ag5275 | |
Primary | |
-20°C | |
CDK6 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CDK6, Cell division protein kinase 6, cyclin dependent kinase 6, PLSTIRE | |
Cdk6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |