Learn More
CDKN1B(phospho T187) & CDKN1B Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Brand:  Abnova DP0086.1Set
Product Code. 16101574
  • £602.00 / 1 set
Explore more special offers
Description
Description
This protein phosphorylation antibody pair set comes with two antibodies, one against the CDKN1B protein, and the other against the specific T187 phosphorylated site of CDKN1B for use in in situ Proximity Ligation Assay.
Specifications
Specifications
CDKN1B
Unconjugated
CDKN4/KIP1/MEN1B/MEN4/P27KIP1
1 Set
Primary
Antibody pair set content: 1. Phospho-CDKN1B T187 rabbit polyclonal antibody (20 ul) With 0.09% sodium azide. 2. CDKN1B mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
In situ PLA
cyclin-dependent kinase inhibitor 1B (p27, Kip1)
CDKN1B
RUO
1027
Human
Ab Pair