missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ CDM™ Chondrocyte Differentiation Medium BulletKit™

Complete kit, basal medium and supplements, to support chondrocyte differentiation.

Brand:  Lonza™ CC-3225

Code : NEW

Additional Details : Weight : 1.00000kg

Product Code. 11605470

  • £720.00 / Each
Currently Unavailable: We have no information when this product will be available
View alternative products

Description

Description

  • Culture kit containing basal medium and supplements.
  • Developed to support the differentiation of chondrocytes.
  • Includes the following:
    • 1 x bottle Basal Medium, 250mL
    • 1 x Yellow Cap Vial with R3-IGF 0.50 mL
    • 1 x Blue Cap Vial with TGF-beta 1.25 mL
    • 1 x Lilac Cap Vial with Insulin 0.50 mL
    • 1 x Red Cap Vial with GA-1000 0.25 mL
    • 1 Bottle FBS 12.50 mL
    • 1 x Natural Cap Vial with Transferrin 0.50 mL
Specifications

Specifications

Medium: Red
Chondrocytes
7.6 to 8.0
Basal Medium
Stem Cell Differentiation Media
Medium: 250 mL, R3-IGF: 0.50 mL, TGF-beta: 1.25 mL, Insulin: 0.50 mL, GA-1000: 0.25 mL, FBS: 12.50 mL, Transferrin: 0.50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers