missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GE Healthcare™ CDM4MAb Liquid Medium without L-glutamine
ADCF medium for use with hybridoma and myeloma cell lines.
Brand: GE Healthcare™ SH30802.LS
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- HyClone CDM4MAb is a chemically defined and animalderived component-free (ADCF) medium.
- This regulatoryfriendly medium is developed through the HyClone Metabolic Pathway Design process to increase process yields for the manufacture of antibodies and antibody fragments for therapeutic use in a variety of engineered hybridoma and myeloma cell lines.
- CDM4MAb medium has been successfully tested in a variety of culture systems, including T-flasks, shaker flasks, and bioreactors including fed-batch and perfusion cultures. CDM4MAb medium does not contain phenol red and is available with or without L-glutamine and poloxamer 188.
- CDM4MAb is available in liquid and powder formats in userfriendly packaging.
Specifications
2° to 8 °C, away from light | |
Liquid | |
6 x 1000 mL |
With: Sodium bicarbonate Without: L-glutamine, poloxamer 188, phenol red, HEPES | |
0.2 μm Sterile-filtered |