missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ CDP-Star™ Substrate (12.5 mM Concentrate)
CDP-Star™ Substrate (12.5 mM Concentrate)
Brand: Molecular Probes™ T2306
Code : 50
Additional Details : Weight : 0.02000kg
Description
This concentrated CDP-Star™ Substrate is a chemiluminescent alkaline phosphatase substrate for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.This CDP-Star™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.
This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours.
For Research Use Only. Not for use in diagnostics procedures.
Specifications
5mL | |
5mL | |
Bottles | |
CDP-Star™ Substrate |
Protein assays and analysis, southern, northern, western blotting, can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays, protein biology, cell analysis, enzyme reporter assays, ELISA, alkaline phosphatase assays, build your own immunoassay, ready-to-use immunoassay | |
AP (Alkaline Phosphatase) Substrate | |
CDP-Star™ Substrate, 5mL, 12.5mM Concentration, Liquid Form, Chemiluminescent Alkaline Phosphatase Substrate For Protein or Nucleic Acid Blotting on Nitrocellulose membranes, Ideal For Protein Assays and Analysis, Southern, Northern and Western blotting, Can also be used in Solution-based Assays | |
4°C |
For Research Use Only. Not for use in diagnostic procedures.