Learn More
Vileda Professional™ CE SoftWall Cleaner
Specialty above-the-floor tools are ideal for surfaces that are difficult to reach, clean and disinfect. Tools are easy to use and ergonomic allowing full range of motion during cleaning and disinfecting
Brand: Vileda Professional™ 162232
Additional Details : Weight : 1.32000kg
Description
- Designed to clean soft wall curtains separating zones within cleanrooms
- Manufactured of electro-polished stainless steel and aluminum
- Completely autoclavable
- Robust contruction and pump action design means higher pressure can be applied during cleaning and easily adjusted as needed
- Unique design allows full contact on curtain from ceiling to floor
Specifications
Curtain Cleaner Tool | |
50 in. | |
Electropolished Stainless Steel, Aluminum |
Non-sterile | |
1 |