Learn More
Vileda Professional™ CE Sweeper and Pan™
The CE Sweeper and Pan™ is ideal for ergonomic and efficient liquid removal. The system is hygienic, lightweight, and flexible. Perfect for dry or damp conditions.
Brand: Vileda Professional™ 119908
105.17 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : Weight : 1.43000kg
Description
- Sweep up broken glass in your cleanroom quickly and easily
- No need to change gloves when an item is dropped on the floor
- Keeps workers from touching the floor within the controlled environment
- Can be sprayed or wiped down with common disinfectants
- Stands up easily for storage or can be hung
- Adjustable bag holder for different size collection bags
Specifications
Dustpan Squeegee Sweeper | |
To sweep broken Glass, for the lobby dustpans and smooth floors where dirt could be dry or damp | |
Janitorial |