Learn More
Vileda Professional™ CE UltraSpeed Pro™ Double Bucket System
The most effective and easy to use flat mop bucket and wringer system for use on floors, walls and ceilings within Controlled Environments!
Brand:  Vileda Professional™ 157775
Product Code. 17882486
  • £275.00 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Demands on effectiveness, cost, ease of handling and staff training is constantly increasing. Whatever the cleaning requirements, the CE UltraSpeed Pro™ Double Bucket System will dramatically increase speed and cleaning performance while decreasing total costs and providing versatility and ergonomics.
  • Double bucket systems includes two buckets with flat mop wringer and chassis
  • Lightweight double bucket system allows for separation of dirty liquid and clean liquid minimizing cross-contamination
  • Touchless wringing of mops using the patented, fast and efficient wringer/press
  • Small and compact chassis (only 2.29 sq ft) means minimal storage space needed
  • Changeable color-coding clips within the buckets (red, blue, yellow, green)
  • Completely autoclavable with all stainless steel parts
  • Highly durable and withstands all common disinfectants
Specifications
Specifications
Autoclavable