missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators Product Code.: 17322743

Thermo Scientific™ CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators

Product Code. 17322743 Shop All Thermo Scientific Products
CsA PLUS Combo Kit
Click to view available options
Type:
CsA PLUS Combo Kit
CsA PLUS High Calibrator Set
This item is not returnable. View return policy

Product Code. 17322743

Brand: Thermo Scientific™ 100147

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Achieve optimal low-end sensitivity and a broad dynamic range with the Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay.

Achieve precise and accurate quantitative results crucial for monitoring drug levels of cyclosporine in kidney, liver and heart transplant patients with our easy-to-use Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay. Used with the Thermo Scientific CEDIA Plus High Range Calibrators, this immunoassay achieves a broad dynamic range of 25-2000ng/mL for both trough and peak samples. Applications are available on our easy-to-use Indiko™ and Indiko Plus clinical and specialty chemistry systems as well as a wide variety of other laboratory instrumentation.

Accurate:
  • Excellent low-end sensitivity
  • Correlation with gold standard methods
  • Precise throughput standard curve assay range
  • Lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Cyclosporine (CsA)
Sample Type Plasma, Serum, Plasma, Serum
Type CsA PLUS Combo Kit
Quantity 41 mL, 19 mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberCOA_100147_ Certificate TypeCertificate of Analysis Date12/02/2025 Product Code,100147
Lot Number75041625_103125 Certificate TypeCertificate of Analysis Date12/02/2025 Product Code,100147
Lot Number74843838_033125 Certificate TypeCertificate of Analysis Date12/02/2025 Product Code,100147
Lot Number74651964 Certificate TypeCertificate of Analysis Date12/02/2025 Product Code,100147
Lot Number74492149 Certificate TypeCertificate of Analysis Date12/02/2025 Product Code,100147
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.