Learn More
Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators
Save time while maintaining accuracy with economical Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators, optimized for use with CEDIA TDM Assays.
£191.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Sample Type | Plasma, Serum |
Type | Calibrator |
Quantity | Low: 2 x 7.5 mL; High: 2 x 5.0 mL |
Product Code | Brand | Description | Detectable Analytes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Detectable Analytes | Price | Quantity & Availability | ||||
17302743
|
Thermo Scientific™
100007 |
CEDIA Antiepileptic TDM Multi-Calibrators | Carbamazepine, Phenobarbital, Phenytoin, Theophylline, Valproic Acid |
£191.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Streamline testing with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators, a single calibrator kit that can be used with multiple CEDIA TDM assays. These economical kits are valued for their accuracy, precision and lot-to-lot dependability. They are available for cardiac or antiarrhythmic, antiepileptic and antibiotic drug monitoring in human serum and plasma. Each kit includes both high and low calibrators.
- Proven technology
- Liquid ready-to-use: no reconstitution needed
- Multiple drug constituents per calibration kit
- Positive linear calibration curve: the higher the absorbance reading, the more drug present
- Excellent correlation to reference methods
- Recombinant DNA technology provides lot-to-lot consistency
Specifications
2°C to 8°C | |
Plasma, Serum | |
Calibrator | |
Low: 2 x 7.5 mL; High: 2 x 5.0 mL |