missing translation for 'onlineSavingsMsg'
Learn More
Cefadroxil, MP Biomedicals™ Product Code.: 11437881

Cefadroxil, MP Biomedicals™

Product Code. 11437881 Shop All MP Biomedicals Products
5 g
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy

Product Code. 11437881

Brand: MP Biomedicals 0215058105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 50370-12-2
Molecular Formula C16H17N3O5S
Molecular Weight (g/mol) 363.388
MDL Number MFCD00865091
InChI Key BOEGTKLJZSQCCD-UEKVPHQBSA-N
Synonym p-hydroxycephalexine, Cefamox
PubChem CID 47965
ChEBI CHEBI:3479
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)SC1)C(=O)O

Specifications

Type Cefadroxil
Physical Form Crystalline Powder
Color White
Quantity 5 g
Formula Weight 363.4
Chemical Name or Material Cefadroxil
compliance-icons compliance-icons
Product Identifier
  • Cefadroxil
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Cefadroxil
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cefadroxil, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.